Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Cerium(III) oxalate nonahydrate ≥99.9% (Ce basis)


93-5832-100G 93-5832-25G
100200-594EA 150.03 USD InStock
100200-594 100197-912
Cerium(III) oxalate nonahydrate ≥99.9% (Ce basis)
Cerium (III) oxalate nonahydrate


Formula: C₆Ce₂O₁₂·9H₂O
Melting Pt: Dec.
MDL Number: MFCD00150140
CAS Number: 13266-83-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty