Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Potassium tetrachloroaurate hydrate ≥99.99% (Au basis) (51%-Au), PURATREM

Warning

93-7906-1G 93-7906-5G
100201-148EA 167.02 USD InStock
100201-148 100201-150
Potassium tetrachloroaurate hydrate ≥99.99% (Au basis) (51%-Au), PURATREM
Potassium tetrachloroaurate hydrate


Formula: AuCl₄K·xH₂O
MDL Number: MFCD00043088
CAS Number: 13682-61-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty