DEV Tryptophan Broth, Dehydrated Culture Media, HiMedia Laboratories
Supplier: PAESEL+LOREI GMBH&CO
GM1901-2.5KG GM1901-500G GM1901-5KG
75804-238EA 739.05 USD InStock
75804-238 75804-240 75804-242
DEV Tryptophan Broth, Dehydrated Culture Media, HiMedia Laboratories
Media for Microbiology Dehydrated Media for Microbiology
DEV tryptophan broth medium can be used for the subcultivation of coliform, differentiation and for indole testing in the bacteriological examination of water.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty