Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Avidin (from Egg white), MP Biomedicals

Supplier: E. Becker & Co. GmbH

0215004710 0215004725 0215004780
76204-016EA 186.25 USD InStock
76204-016 76204-018 76204-020
Avidin (from Egg white), MP Biomedicals
Proteins and Peptides
Avidin is a basic glycoprotein isolated from raw egg white. It exhibits high binding affinity for biotin and is capable of producing biotin deficiency in rats and chicks. It also occurs in the white portion of eggs and the oviducts of birds and amphibia. It is destroyed by cooking or irradiation.

  • Ready for CE IVD labeling of clinical applications
  • Specific activity: 10 to 15 u/mg solid
  • Solubility: Soluble in water or dilute aqueous buffer
  • Unit definition: One unit will bind 1 µg of D-biotin at pH 8.9

Avidin is useful in a biotin-avidin system for labeling biomolecules for receptor studies, immunoassays and immunohistological methods. Avidin egg white was used in an assay using functionalized xenon as a biosensor to detect biotin-avidin binding. Egg white was used at 80 nmol. Egg white avidin has been used as a cross-linker for biotin-conjugated antibodies. Avidin also been used as a blocking agent in ELISA applications.

Precaución: Research use only (RUO).


Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty