Powder-Free Nitrile Decontamination Exam Gloves, Cardinal
Supplier: Cardinal Health
88NDS 88NDM 88NDL 88NDXL
76264-574CS 570.88 USD InStock
76264-574 76264-576 76264-578 76264-580
Powder-Free Nitrile Decontamination Exam Gloves, Cardinal
Gloves
Nitrile Decontamination exam gloves feature a 16" cuff to provide added coverage and protection.
  • Thickest Cardinal Health® nitrile exam glove in the portfolio
  • 16" extended length
  • Full textured fingers
  • Tested for use with various lab chemicals and chemotherapy drugs
  • Tested for puncture resistance
Nitrile Decontamination is the thickest glove in the Cardinal Health® synthetic exam glove portfolio, designed to offer protection where there is potential exposure to sharps and chemicals.
Certificaciones: ASTM D6978, ASTM F1342-91.
Envase: Packaged 50 gloves per box, 10 boxes per case.
Precaución: Gloves used for protection against laboratory chemical exposure should be selected specifically for the type of chemicals being used. Review Safety Data Sheets (SDS) for the chemicals/drugs being used to determine required level of protection. This glove has been tested for resistance to permeation of various chemotherapy drugs per ASTM D6978, 'Standard Practice for Assessment of Resistance of Medical Gloves to Permeation by Chemotherapy Drugs'. This glove has been tested for puncture resistance per ASTM F132-91, 'Standard Test Method for Protective Clothing Material Resistance to Puncture'.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty