UniStore™ Reagent Bottles, Wide Mouth, HDPE
Supplier: United Scientific Supplies
33406 33407 33408 33409 33410
76263-098PK 22.88 USD InStock
76263-098 76263-100 76263-102 76263-104 76263-106
UniStore™ Reagent Bottles, Wide Mouth, HDPE
Bottles Plastic Bottles
Bottles are for general laboratory use and for storing, shipping, and packing liquids.
  • Translucent
  • Pliable
UniStore™ wide mouth HDPE reagent bottles are manufactured from USP Class VI resin that is FDA-21CFR compliant. Excellent chemical resistance to most acids, bases, and alcohols.
Información para pedidos: Polypropylene caps are included.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty