Separatory Funnels, United Scientific Supplies
Supplier: United Scientific Supplies
81201-VWR 81202 81203
76263-254EA 31.4 USD InStock
76263-254 76263-256 76263-258
Separatory Funnels, United Scientific Supplies
Funnels Separating Funnels
Resists virtually all chemicals.
  • Stem length: 45 mm
  • Includes a leak-proof threaded screw cap
  • Funnels are not graduated
These pear-shaped polypropylene separating funnels feature excellent clarity and strength. The polypropylene stopcock is fitted with a self-lubricating PTFE plug.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty