Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Anti-CLEC9A Monoclonal Antibody

Supplier: Bio X Cell
BE0305-100MG BE0305-5MG BE0305-25MG BE0305-1MG BE0305-50MG
103572-678EA 3920 USD InStock
103572-678 103572-680 103572-682 103572-684 103572-686 76554-780 76555-264 76556-154 76556-566 76556-940
Anti-CLEC9A Monoclonal Antibody
Antibodies
InVivoMAb anti-mouse CLEC9A (CD370), Clone: 7H11, Isotype: Rat IgG1, κ, Applications: in vivo Ag targeting to CLEC9A+ DCs, Western blot, ELISA, Immunoprecipitation, Immunofluorescence, Flow cytometry

Type: Primary
Antigen: CLEC9A
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope: Rat
Host:
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty