Mouse Recombinant TFF-2 (from E. coli)
Supplier: PEPROTECH INC.
315-30-5UG 315-30-20UG 315-30-100UG 315-30-250UG 315-30-500UG 315-30-1MG
76077-450EA 132.81 USD InStock
76077-450 76077-452 76077-454 76077-456 76077-458 76077-460
Mouse Recombinant TFF-2 (from E. coli)
Proteins and Peptides
The Trefoil Factor peptides (TFF1, TFF2 and TFF3) are expressed in the gastrointestinal tract, and appear to play an important role in intestinal mucosal defense and repair. TFF2 has been shown to inhibit gastrointestinal motility and gastric acid secretion. Recent data suggests a potential role for TFF2 in acute and chronic asthma (Nikolaidis, N.M. et al. Am. Journal Respir. Cell Mol. Biol. (2003) 4: 458-464). Recombinant Murine TFF-2 is an 11.9 kDa polypeptide of 106 amino acid residues, which includes a 40-amino acid trefoil motif containing three conserved intramolecular disulfide bonds.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty