Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Anti-GRO-beta/MIP-2 (CXCL2) Rabbit Polyclonal Antibody

Supplier: PeproTech, Inc.
500-P75-50UG 500-P75-100UG 500-P75-1MG
10775-678EA 339.8 USD InStock
10775-678 10775-680 10775-682
Anti-GRO-beta/MIP-2 (CXCL2) Rabbit Polyclonal Antibody
Antibodies
Produced from sera of rabbits immunized with highly pure Recombinant Rat GRO-β/MIP-2 (CXCL2). Anti­Rat GRO-β/MIP-2 (CXCL2)­specific antibody was purified by affinity chromatography employing an immobilized Rat GRO-β/MIP-2 matrix.

Applications tested can vary by lot number. See Product Data Sheet for more information.

Type: Primary
Antigen: GRO-beta/MIP-2 (CXCL2)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Rat

Order Now
 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty