LR3IGF-I Rapid ELISA Kit: Human, Biosensis
Supplier: ACS Scientific Inc
BEK-2233-1P BEK-2233-2P
76264-590EA 437.85 USD InStock
76264-590 76264-592
LR3IGF-I Rapid ELISA Kit: Human, Biosensis
Assays ELISAs
The ELISA kit consists of a complete set of reagents and pre-coated plate to allow immediate assay of LR³IGF-I in culture media. Included in the kit are mouse monoclonal anti- LR³IGF-I capture antibody pre-coated onto an ELISA plate, standard LR³IGF-I protein, a biotinylated anti- LR³IGF-I detection antibody and horseradish peroxidase (HRP)-conjugated streptavidin.
The addition of a substrate (3,3’,5,5’-tetramethylbenzidine, TMB) yields a colored reaction product which is directly proportional to the concentration of LR³IGF-I present in samples and the supplied protein standard. This LR³IGF-I RapidTM ELISA kit has been developed, optimized and validated to quantify LR³IGF-I protein in cell culture medium.
It is likely to be used to measure LR³IGF-I in media and during downstream processing of media following a production cycle and is not intended for other use.
Información de suministro: The ELISA kit box contains 1 or 2 x 96-well pre-coated strip plate, protein standards, detection reagents, wash and sample buffers, substrate buffer and detailed protocols.
Precaución: This kit has been configured for research use only and is not to be used in diagnostic or clinical procedures.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty