Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Galantamine hydrobromide

Danger

H37491-03 H37491-06
AAH37491-03EA 416.99 USD InStock
AAH37491-03 AAH37491-06
Galantamine hydrobromide
Galantamine hydrobromide


Formula: C₁₇H₂₁NO₃·HBr
Storage Temperature: Ambient
MDL Number: MFCD00067672
CAS Number: 1953-04-4
EINECS: 217-780-5
UN: 2811
ADR: 6.1,III
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty