Tech-Med® Stainless Steel Dressing Jars, DUKAL Corporation®
Supplier: Dukal
4233 4233-1 4233-2
76235-298BX 849.71 USD InStock
76235-298 76235-464 76235-466
Tech-Med® Stainless Steel Dressing Jars, DUKAL Corporation®
Jars General Use Jars
Stainless steel jars are crafted without seams to prevent bacteria growth and allow for easy cleaning.
  • Made of the highest grade stainless steel (18/8)
  • Autoclavable
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty