Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Anti-KDEL Mouse Monoclonal Antibody [clone: 10C3]

Supplier: Enzo Life Sciences
ABS679-0050 ABS679-0200
76271-442EA 359.7 USD InStock
76271-442 76271-444
Anti-KDEL Mouse Monoclonal Antibody [clone: 10C3]
Antibodies
Recommended Applications: ELISA, Flow Cytometry, ICC, IF, IHC (PS), IP, WB, Electron microscopy

Type: Primary
Antigen: KDEL
Clonality: Monoclonal
Clone: 10C3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Yeast;Plant;Mammals;Insect;Avian

Order Now
 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty