Magid® MultiMaster® Nitrile Palm Coated Gloves, Magid
Supplier: Magid Glove
N95OE N95OEC
69100-391PK 55.8 USD InStock
69100-391 69100-393
Magid® MultiMaster® Nitrile Palm Coated Gloves, Magid
Gloves
Magid's MultiMaster glove is a comfortable, seamless machine knit glove that a features a blue double nitrile palm coating.
  • Double nitrile palm-coating provides firm grip, excellent durability
  • Cotton/polyester shell ensures comfort
  • Hemmed edge prevents unraveling in the wash
  • Knit wrist provides a snug fit
The nitrile palm coating offers a rock-solid grip and excellent flexibility, great for multi-purpose and general handling protection.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty