Platinum Palladium (80:20), sputtering target sputter coater cathodes
Supplier: EPREDIA
91006-PP 91007-PP 91008-PP 91009-PP 91010-PP 91013-PP 91014-PP 91015-PP 91016-PP 91017-PP 91018-PP 91019-PP 91020-PP
102099-414EA 808.58 USD InStock
102099-414 102099-444 102099-474 102099-504 102099-532 102099-564 102099-594 102099-624 102099-654 102099-684 102099-714 102099-744 102099-774
Platinum Palladium (80:20), sputtering target sputter coater cathodes
Platinum Palladium
EMS (ELECTRON MICROSCOPY SCIENCES) precious metal sputtering targets are made from high purity metals, starting 99.99% with most standard disc sizes, for use on most sputtering units from many manufacturers: Emitech, Emscope, Bio-Rad, Polaron, Edwards, Balzers, Plasma Sciences, Technics Hummers, Denton, Cressington, and much more.
EMS targets come in a standard thickness of 0.1 mm (100µm). Other thickness, ranging from 0.05 to 6mm, are available upon request. Targets are avaible mounted or unmounted whichever you prefer. For mounting please always state the make and the model of the sputtering unit so we can tend to it for you. Targets in other materials such as Chromium, Iridium etc. are available upon request. We also can manufacture targets with diameters from 4 mm to 304.8 mm (12"). Call for pricing.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty