Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Nitric acid 5% (v/v)

Supplier: RICCA CHEMICAL CO
Danger

R5318000-1A R5318000-20F R5318000-4A R5318000-500A
RCR5318000-1AEA 93 USD InStock
RCR5318000-1A RCR5318000-20F RCR5318000-4A RCR5318000500A
Nitric acid 5% (v/v)
Nitric acid


Formula: HNO₃
MW: 63.01
Boiling Pt: 87
Melting Pt: -42
Density: 1.51
Storage Temperature: Ambient
MDL Number: MFCD00011349
CAS Number: 7697-37-2
UN: 3264
ADR: 8 III
Merck Index: 13,06610
Order Now
Specification Test Results
Appearance Colorless liquid
Assay (vs. Sodium Hydroxide/Phenolphthalein) 4.99-5.01 % (v/v)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty