LB Agar Medium, MP Biomedicals
Supplier: E. Becker & Co. GmbH
113002232 113002242
76020-262EA 227.56 USD InStock
76020-262 IC113002242
LB Agar Medium, MP Biomedicals
Cell Culture Media
Rich medium for the growth of E. coli and certified for plasmid and other bacterial cultures in liquid medium.
  • Proprietary media for enhanced cell yield
  • Convenient packaging formats for a variety of applications and budgets
  • Proven and reliable reputation
  • High-quality raw materials and additives
LB Agar Medium is a nutritionally rich medium originally developed for growth and maintenance of E. coli strains. LB agar medium supports a luxurious growth of E. coli cells as tryptone and yeast extract supply essential growth factors that E. coli cells would otherwise have to synthesize. The medium also contains essential electrolytes for transport and osmotic balance, due to the NaCl component.
Application includes: Bacteria cell culture, gene expression, transformation, microbiological analysis, protein expression.
Please note that the pH of LB Agar Medium increases as the temperature of the solution decreases.
Certificaciones: Molecular Biology Certified™.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty