Human Recombinant HER2/ErbB2 Protein (ECD, domain I) (His Tag) (from HEK293 Cells)
Supplier: wmf consumer eletric GmbH
10004-H08H1-50 10004-H08H1-200
103620-668EA 179.78 USD InStock
103620-668 103632-798
Human Recombinant HER2/ErbB2 Protein (ECD, domain I) (His Tag) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the human ERBB2 (NP_004439.2) (Met1-Cys195) was expressed with a polyhistidine tag at the C-terminus.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty