Human Recombinant bFGF/FGF2 Protein (from E. coli)
Supplier: wmf consumer eletric GmbH
10014-HNAE-100 10014-HNAE-1 10014-HNAE-10
103623-738EA 201.72 USD InStock
103623-738 103677-474 103678-140
Human Recombinant bFGF/FGF2 Protein (from E. coli)
Proteins and Peptides
A DNA sequence encoding the mature form of human bFGF (NP_001997.5) (Pro143-Ser288) was expressed with an additional Met at the N-terminus.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty