Human Recombinant IL33 (from E. coli)
Supplier: wmf consumer eletric GmbH
10368-HNAE-100 10368-HNAE-5
103621-066EA 452.48 USD InStock
103621-066 103677-498 103679-094
Human Recombinant IL33 (from E. coli)
Proteins and Peptides
A DNA sequence encoding the C-terminal segment of human IL33 (NP_254274.1) (Ser 112-Thr 270) was expressed with an initial Met at the N-terminus.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty