Human/Rhesus/Cynomolgus/Canine Recombinant TGF-beta 1/TGFB1 Protein (from CHO cells)
Supplier: wmf consumer eletric GmbH
10804-HNAC-500 10804-HNAC-100
103622-552EA 2179.58 USD InStock
103622-552 103622-554 103677-866
Human/Rhesus/Cynomolgus/Canine Recombinant TGF-beta 1/TGFB1 Protein (from CHO cells)
Proteins and Peptides
A DNA sequence encoding the active form of human / rhesus / cynomolgus / canine TGFβ1 (NP_000651.3) (Ala 279-Ser 390) was expressed and purified. human, Rhesus, cynomolgus and Canine TGFβ1 sequences are identical.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty