C. Perfringens Recombinant Neuraminidase/NA (Active) (His Tag) (from E. coli)
Supplier: wmf consumer eletric GmbH
11680-V07E-10 11680-V07E-20
103612-552EA 179.78 USD InStock
103612-552 103612-554
C. Perfringens Recombinant Neuraminidase/NA (Active) (His Tag) (from E. coli)
Proteins and Peptides
A DNA sequence encoding the C. Perfringens neuraminidase (P10481.1) (Cys 2-Gln 382) was expressed, with a N-terminal polyhistidine tag.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty