Human Recombinant IL4/Interleukin-4 Protein (from E. coli)
Supplier: wmf consumer eletric GmbH
11846-HNAE-100 11846-HNAE-1
103622-832EA 454.88 USD InStock
103622-832 103677-642 103677-644
Human Recombinant IL4/Interleukin-4 Protein (from E. coli)
Proteins and Peptides
A DNA sequence encoding the mature form of human IL4 isoform 1 (P05112-1) (His 25-Ser 153) was expressed and purified with an initial Met.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty