Mouse Recombinant IL21/Interleukin 21 Protein (from E. coli)
Supplier: wmf consumer eletric GmbH
50137-MNAE-100 50137-MNAE-20
103664-656EA 448 USD InStock
103664-656 103666-862 103678-612
Mouse Recombinant IL21/Interleukin 21 Protein (from E. coli)
Proteins and Peptides
A DNA sequence encoding the soluble form of mouse IL21 (Q9ES17) (Pro 25-Ser 146) was expressed, with an initial Met at the N-terminus.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty