Mouse Recombinant β-NGF/Beta-NGF Protein (from CHO cells)
Supplier: wmf consumer eletric GmbH
50385-MNAC-100 50385-MNAC-20 50385-MNAC-1
103664-940EA 300.98 USD InStock
103664-940 103664-942 103667-326 103677-824 103677-826
Mouse Recombinant β-NGF/Beta-NGF Protein (from CHO cells)
Proteins and Peptides
A DNA sequence encoding the mature form of mouse NGF (NP_001106168.1) (Ser 122-Gly 241) was expressed, with an initial Met at the C-terminus.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty