Graham Condensers, 24/40 Joint, United Scientific Supplies
Supplier: United Scientific Supplies
CNG200-VWR CNG300-VWR CNG500-VWR
76293-450EA 74.9 USD InStock
76293-450 76293-452 76293-456
Graham Condensers, 24/40 Joint, United Scientific Supplies
Condensers
Graham condensers are designed for use in distillation applications.
  • 24/40 sockets
  • Screw thread connector
Constructed of borosilicate glass, this condenser has a coiled inner tube to provide additional surface area for highly efficient cooling. Socket is 24/40 with a screw thread connector.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty