Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Anti-Myc tag Mouse Monoclonal Antibody [clone: 08]

Supplier: Sino Biological
100029-MM08-100 100029-MM08-500
103589-986EA 164.32 USD InStock
103589-986 103628-446 103628-448 103638-040 103638-042
Anti-Myc tag Mouse Monoclonal Antibody [clone: 08]
Antibodies
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, a synthetic Myc tag peptide conjugated to KLH. The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.

Type: Primary
Antigen: Myc tag
Clonality: Monoclonal
Clone: 08
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: Mouse IgG1
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty