Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Anti-BCL2 Rabbit Monoclonal Antibody [clone: 204] (PerCP (Peridinin-Chlorophyll Protein Complex))

Supplier: Sino Biological
100126-R204-C-25 100126-R204-C-100
103630-342EA 90.9 USD InStock
103630-342 103632-528
Anti-BCL2 Rabbit Monoclonal Antibody [clone: 204] (PerCP (Peridinin-Chlorophyll Protein Complex))
Antibodies
This antibody was obtained from a rabbit immunized with a synthetic peptide corresponding to the N-terminus of the Human BCL2/Bcl-2, and conjugated with PerCP under optimum conditions, the unreacted PerCP was removed.

Type: Primary
Antigen: Bcl-2
Clonality: Monoclonal
Clone: Clone: 204
Conjugation: PerCP (Peridinin-Chlorophyll Protein Complex)
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty