Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Anti-TLR2 Rabbit Monoclonal Antibody [clone: 067] (PE (Phycoerythrin))

Supplier: Sino Biological
10061-R067-P-25 10061-R067-P-100
103628-732EA 90.9 USD InStock
103628-732 103630-410
Anti-TLR2 Rabbit Monoclonal Antibody [clone: 067] (PE (Phycoerythrin))
Antibodies
This antibody was obtained from a rabbit immunized with purified, recombinant Human TLR2 / CD282 (rh TLR2 / CD282; Catalog#10061-H08B1; NP_003255.2; Met1-Arg587) and conjugated with PE under optimum conditions, the unreacted PE was removed.

Type: Primary
Antigen: TLR2
Clonality: Monoclonal
Clone: Clone: 067
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty