Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Anti-FCGR2A Mouse Monoclonal Antibody [clone: 01] (FITC (Fluorescein Isothiocyanate))

Supplier: Sino Biological
10374-MM01-F-25 10374-MM01-F-100
103628-986EA 70.7 USD InStock
103628-986 103631-564
Anti-FCGR2A Mouse Monoclonal Antibody [clone: 01] (FITC (Fluorescein Isothiocyanate))
Antibodies
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human CD32a/FCGR2A/Fc gamma RIIA (rh CD32a/FCGR2A/Fc gamma RIIA; Catalog#10374-H08H; P12318-1; Met1-Ile218,167R) and conjugated with FITC under optimum conditions, the unreacted FITC was removed.

Type: Primary
Antigen: CD32a
Clonality: Monoclonal
Clone: Clone: 01
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype: Mouse IgG1
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty