Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Anti-DPP4 Mouse Monoclonal Antibody [clone: 05] (APC (Allophycocyanin))

Supplier: Sino Biological
10688-MM05-A-100 10688-MM05-A-25
103630-746EA 222.2 USD InStock
103630-746 103631-342
Anti-DPP4 Mouse Monoclonal Antibody [clone: 05] (APC (Allophycocyanin))
Antibodies
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human DPP4 / CD26 (rh DPP4 / CD26; Catalog#10688-HNCH; NP_001926.2; Asn29-Pro766) and conjugated with APC under optimum conditions, the unreacted APC was removed.

Type: Primary
Antigen: CD26
Clonality: Monoclonal
Clone: Clone: 05
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: Mouse IgG1
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty