Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Anti-MET Rabbit Monoclonal Antibody [clone: 271] (APC (Allophycocyanin))

Supplier: Sino Biological
10692-R271-A-25 10692-R271-A-100
103629-130EA 70.7 USD InStock
103629-130 103631-582
Anti-MET Rabbit Monoclonal Antibody [clone: 271] (APC (Allophycocyanin))
Antibodies
This antibody was obtained from a rabbit immunized with purified, recombinant Human HGFR / c-MET (rh HGFR / c-MET; Catalog#10692-H08H; NP_000236.2; Met 1-Thr 932) and conjugated with APC under optimum conditions, the unreacted APC was removed.

Type: Primary
Antigen: c-MET
Clonality: Monoclonal
Clone: Clone: 271
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty