Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Anti-TNFRSF1A Rabbit Monoclonal Antibody [clone: R111]

Supplier: Sino Biological
10872-R111-500 10872-R111-200
103618-154EA 403.2 USD InStock
103618-154 103618-156
Anti-TNFRSF1A Rabbit Monoclonal Antibody [clone: R111]
Antibodies
This antibody was obtained from a rabbit immunized with purified, recombinant Human TNFR1 / CD120a / TNFRSF1A (rh TNFR1 / CD120a / TNFRSF1A; Catalog#10872-H08H; NP_001056.1; Met1-Thr211) and was produced using recombinant antibody technology.

Type: Primary
Antigen: TNFR1
Clonality: Monoclonal
Clone: R111
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty