Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Anti-RSV-F Rabbit Monoclonal Antibody [clone: R338]

Supplier: Sino Biological
11049-R338-200 11049-R338-500
103619-132EA 252 USD InStock
103619-132 103619-134
Anti-RSV-F Rabbit Monoclonal Antibody [clone: R338]
Antibodies
This antibody was obtained from a rabbit immunized with purified, recombinant Human respiratory syncytial virus (RSV) Fusion glycoprotein / RSV-F (Catalog#11049-V08B; P03420.1; Met1-Thr529) and was produced using recombinant antibody technology.

Type: Primary
Antigen: RSV-F
Clonality: Monoclonal
Clone: R338
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty