Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Anti-CD28 Rabbit Monoclonal Antibody [clone:007] FITC (Fluorescein Isothiocyanate)

Supplier: Sino Biological
11524-R007-F-25 11524-R007-F-100
103629-568EA 30.3 USD InStock
103629-568 103631-642
Anti-CD28 Rabbit Monoclonal Antibody [clone:007] FITC (Fluorescein Isothiocyanate)
Antibodies
This antibody was obtained from a rabbit immunized with purified, recombinant Human CD28 (rh CD28; Catalog#11524-H08H; P10747-1; Met1-Pro152) and conjugated with FITC under optimum conditions, the unreacted FITC was removed.

Type: Primary
Antigen: CD28
Clonality: Monoclonal
Clone: Clone: 007
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty