Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Anti-IL17A Mouse Monoclonal Antibody [clone:M237]

Supplier: Sino Biological
12047-M237-200 12047-M237-500
103636-268EA 252.5 USD InStock
103636-268 103636-270
Anti-IL17A Mouse Monoclonal Antibody [clone:M237]
Antibodies
This antibody was obtained from a mouse immunized with purified, recombinant Human IL17 / IL17a (rHuman IL17 / IL17a; Catalog#12047-HNAE; Q16552; Ile20-Ala155) and was produced using recombinant antibody technology.

Type: Primary
Antigen: IL17/IL17a
Clonality: Monoclonal
Clone: M237
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: Mouse IgG1
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty