Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Anti-CD58 Rabbit Monoclonal Antibody [clone:083] PerCP (Peridinin-Chlorophyll Protein Complex)

Supplier: Sino Biological
12409-R083-C-25 12409-R083-C-100
103629-778EA 90.9 USD InStock
103629-778 103631-994
Anti-CD58 Rabbit Monoclonal Antibody [clone:083] PerCP (Peridinin-Chlorophyll Protein Complex)
Antibodies
This antibody was obtained from a rabbit immunized with purified, recombinant Human LFA-3 / CD58 (rh LFA-3 / CD58; Catalog#12409-H08H; AAH05930.1; Met1-Arg215) and conjugated with PerCP under optimum conditions, the unreacted PerCP was removed.

Type: Primary
Antigen: CD58
Clonality: Monoclonal
Clone: Clone: 083
Conjugation: PerCP (Peridinin-Chlorophyll Protein Complex)
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty