Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Anti-ALCAM Rabbit Monoclonal Antibody [clone: 018] PE (Phycoerythrin)

Supplier: Sino Biological
90227-R018-P-25 90227-R018-P-100
103630-276EA 90.9 USD InStock
103630-276 103632-472
Anti-ALCAM Rabbit Monoclonal Antibody [clone: 018] PE (Phycoerythrin)
Antibodies
This antibody was obtained from a rabbit immunized with purified, recombinant Cynomolgus CD166/ALCAM ( Catalog#90227-C08H; G7NZQ8; Met1-Ala526) and conjugated with PE under optimum conditions, the unreacted PE was removed.

Type: Primary
Antigen: ALCAM
Clonality: Monoclonal
Clone: Clone: 018
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Cynomolgus

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty