Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Anti-IL12B and IL23A Mouse Monoclonal Antibody [clone: M066]

Supplier: Sino Biological
CT035-M066-500 CT035-M066-200
103662-646EA 404 USD InStock
103662-646 103662-648
Anti-IL12B and IL23A Mouse Monoclonal Antibody [clone: M066]
Antibodies
This product is a recombinant monoclonal antibody expressed from HEK293 cells.

Type: Primary
Antigen: IL23A & IL12B Heterodimer
Clonality: Monoclonal
Clone: M066
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: Mouse IgG1
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty