Human Recombinant Galectin-1
Supplier: Bandelin Electronic GmbH &Co.KG
009-001-U91-0010 009-001-U91-0050
10801-524EA 2179 USD InStock
10801-524 10801-526 10801-528
Human Recombinant Galectin-1
Proteins and Peptides
Recombinant Human Galectin-1 control protein
  • Control Protein
  • PCA Assays
  • ImmunoBlot
Galectin-1 belongs to a large family of carbohydrate-binding proteins called lectins. Galectin-1can be either monomeric or homodimeric and is found in a wide variety of cells and tissue types. Galectin-1 can control cell growth, proliferation, induce apoptosis of activated T cells while it can also modulate cytokine secretion or inhibit pro-inflammatory cytokine production. Galectin-1 plays an important role in acute and chronic inflammation. Recombinant human Galectin-1 is a non-glycosylated protein, containing134 amino acids, with a molecular weight of 14.5 kDa.
Galectin-1 Recombinant Protein is suitable as a control for polyclonal or monoclonal anti-Galectin-1 in immunological assays.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty