Human Recombinant Gro-Beta /CXCL2
Supplier: Bandelin Electronic GmbH &Co.KG
009-001-V16-0010 009-001-V16-0100 009-001-V16-0002
75928-256EA 352.09 USD InStock
75928-256 75928-258 75928-260 75928-646
Human Recombinant Gro-Beta /CXCL2
Proteins and Peptides
Recombinant Human Gro-Beta /CXCL2 control protein
  • Control Protein
  • PCA Assays
  • ImmunoBlot
Three human MGSA/GRO genes encode 3 highly related chemokines, MGSA/GRO α, -β and -g. All 3 MGSA/GRO proteins bind to the same receptors, but with differing affinities, and stimulate a number of biological responses including chemotaxis, angiogenesis, and growth regulation. Recombinant human GROβ is a non-glycosylated protein, containing 73 amino acids, with a molecular weight of 7.9 kDa.
Gro-beta Recombinant Protein is suitable as a control for polyclonal or monoclonal anti-Gro-beta in immunological assays.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty