Human Recombinant Gro-gamma/MIP-2Beta
Supplier: Bandelin Electronic GmbH &Co.KG
009-001-V17-0010 009-001-V17-0100
75928-630EA 4839 USD InStock
75928-630 75928-632 75928-634 75928-636
Human Recombinant Gro-gamma/MIP-2Beta
Proteins and Peptides
Recombinant Human Gro-gamma/MIP-2Beta control protein
  • Control Protein
  • PCA Assays
  • ImmunoBlot
Growth Regulated Proteins (GRO) are a group of three proteins, GRO-α, -β and –g, that are encoded by three distinct genes. All 3 GRO proteins can bind to the same receptors, but with differing affinities, and stimulate a number of biological responses including chemotaxis, angiogenesis, and growth regulation. More specifically, GRO gamma (also called CXCL3), can act through chemokine receptor CXCR2 to promote monocyte migration and adhesion. Recombinant human GRO gamma is a non-glycosylated protein, containing 73 amino acids, with a molecular weight of 7.9 kDa.
GRO-gamma Recombinant Protein is suitable as a control for polyclonal or monoclonal anti-GRO-gamma in immunological assays.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty