Human Recombinant IGF-I
Supplier: Bandelin Electronic GmbH &Co.KG
009-001-V21-0100 009-001-V21-0020
75928-946EA 492 USD InStock
75928-946 75934-652 75934-654
Human Recombinant IGF-I
Proteins and Peptides
Recombinant Human IGF-I control protein
  • Control Protein
  • PCA Assays
  • ImmunoBlot
Insulin-like Growth Factor I, IGF-I, is a growth factor produced by the liver when stimulated with growth hormone and can be found circulating throughout the body . IGF-I activates the IGF-I receptor (IGF1R) and the insulin receptor to mediate growth of almost every cell of the body. IGF-I is known as one of the most potent activators of the AKT signaling pathway which is known to be a stimulator of proliferation and an inhibitor of programmed cell death. Mature human IGF-I is 100% homologous with bovine and porcine proteins. Recombinant human IGF-I is a non-glycosylated protein, containing 70 amino acids, with a molecular weight of 7.7 kDa.
Insulin-like Growth Factor I Recombinant Protein is suitable as a control for polyclonal or monoclonal anti-Insulin-like Growth Factor I in immunological assays.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty