Human Recombinant SDF-1 alpha/CXCL12
Supplier: Bandelin Electronic GmbH &Co.KG
009-001-W22-0010 009-001-W22-0100
75932-328EA 4839 USD InStock
75932-328 75932-332 75932-336 75932-340
Human Recombinant SDF-1 alpha/CXCL12
Proteins and Peptides
Recombinant Human SDF-1 alpha/CXCL12 control protein
  • Control Protein
  • PCA Assays
  • ImmunoBlot
Stromal cell Derived Factor-1 alpha (SDF-1α), also called CXCL12, is one of two splice variants made by a wide variety of cells when stimulated by inflammatory cytokines such as, TNF, IL-1 or LPS. SDF-1α signals through the G protein-couple receptor, CXCR4, to recruit activated leukocytes. Human and mouse SDF-1α share 99% sequence identity. Recombinant human SDF-1α is a non-glycosylated protein, containing 68 amino acids, with a molecular weight of 8 kDa.
Stromal Cell-Derived Factor-1 alpha Recombinant Protein is suitable as a control for polyclonal or monoclonal anti-Stromal Cell-Derived Factor-1 alpha in immunological assays.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty