Human Recombinant Visfatin
Supplier: Bandelin Electronic GmbH &Co.KG
009-001-W40-0100 009-001-W40-0025 009-001-W40-0005
75933-572EA 4839 USD InStock
75933-572 75933-574 75933-576 75933-578
Human Recombinant Visfatin
Proteins and Peptides
Recombinant Human Visfatin control protein
  • Control Protein
  • PCA Assays
  • ImmunoBlot
Visfatin is an adipokine produced by visceral adipose tissue. Other adipokines include leptin, adiponectin, resistin, chemerin, omentin and vaspin. Visfatin has been shown to act as a pro-inflammatory factor for PBMCs and may mimic the effects of insulin on adipocytes, monocytes and hepatocytes. Recombinant human Visfatin is a non-glycosylated protein, containing 466 amino acids, with a molecular weight of 52.6 kDa.
Visfatin Recombinant Protein is suitable as a control for polyclonal or monoclonal anti-Visfatin in immunological assays.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty