Human Recombinant IL-6
Supplier: Bandelin Electronic GmbH &Co.KG
009-F01-310-0100 009-F01-310-0020 009-F01-310-0005
75929-334EA 5322 USD InStock
75929-334 75929-340 75929-346 75934-780
Human Recombinant IL-6
Proteins and Peptides
Recombinant Human IL-6 control protein
  • Control Protein
  • PCA Assays
  • ImmunoBlot
Interleukin 6 (IL-6) is an important pro-inflammatory and anti-inflammatory cytokine expressed by T cells, macrophages and muscle cells. IL-6 signals through a receptor complex containing two receptors, IL-6Rα and gp130. IL-6 has an important function in promoting fever and can serve to stimulate an immune response to trauma. IL-6 is often used for growth of hybridoma cell lines. Human IL-6 is active on mouse and rat cells. Recombinant human IL-6 is a non-glycosylated protein, containing 184 amino acids, with a molecular weight of 21 kDa.
Interleukin-6 Recombinant Protein is suitable as a control for polyclonal or monoclonal anti-Interleukin-6 in immunological assays. Buffer Formulation: 10 mM sodium phosphate, pH 7.5.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty