Human Recombinant FGF147 basic
Supplier: Bandelin Electronic GmbH &Co.KG
009-F01-U82-0010 009-F01-U82-0050
10802-812EA 2384 USD InStock
10802-812 10802-814 10802-816
Human Recombinant FGF147 basic
Proteins and Peptides
Recombinant Human FGF147 basic control protein
  • Control Protein
  • PCA Assays
  • ImmunoBlot
Fibroblast Growth Factors (FGFs) are a 22 member family of proteins known to be involved in angiogenesis, wound healing and embryonic development. As a family, they bind to heparin and signal through four receptor tyrosine kinases called, FGFR1, 2, 3 and 4. Although the mechanism remains unclear, FGF-basic 147 (variant of FGF basic 154), also called FGF-2, is a critical component in keeping embryonic stem cells undifferentiated in cell culture systems. Recombinant human FGF-b 147 (FGF-2) is a non-glycosylated protein, containing 147 amino acids, with a molecular weight of 16.5 kDa.
Fibroblast Growth Factor 147 basic Recombinant Protein is suitable as a control for polyclonal or monoclonal anti-Fibroblast Growth Factor 147 basic in immunological assays.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty