Human Recombinant Flt-3 Ligand
Supplier: Bandelin Electronic GmbH &Co.KG
009-F01-U86-0010 009-F01-U86-0100
10802-904EA 6969 USD InStock
10802-904 10802-906 10802-908 10802-910
Human Recombinant Flt-3 Ligand
Proteins and Peptides
Recombinant Human Flt-3 Ligand control protein
  • Control Protein
  • PCA Assays
  • ImmunoBlot
FMS-related Tyrosine Kinase 3 Ligand (FLT-3 Ligand) is a growth factor important for the proliferation of hematopoietic cells. FLT-3 Ligand binds to, and transmits signals through, the receptor tyrosine kinase known as FMS-like Tyrosine Kinase-3 (FLT-3). FLT-3 Ligand promotes long-term expansion and differentiation of human pro-B cells in the presence of IL-7 or in combination of IL-7 and IL-3. Human FLT-3 Ligand can stimulate the proliferation of cells expressing murine FLT-3 receptors. Recombinant human FLT-3 Ligand is a non-glycosylated protein, containing 155 amino acids, with a molecular weight of 17.6 kDa.
Flt-3 Ligand Recombinant Protein is suitable as a control for polyclonal or monoclonal anti- Flt-3 Ligand in immunological assays. Buffer Formulation: 10 mM sodium phosphate and 50 mM sodium chloride, pH 7.5.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty