Human Recombinant NGF beta (from E. coli)
Supplier: Bandelin Electronic GmbH &Co.KG
009-F01-W03-0100 009-F01-W03-0020 009-F01-W03-0005
75930-268EA 5322 USD InStock
75930-268 75930-270 75930-272 75930-274
Human Recombinant NGF beta (from E. coli)
Proteins and Peptides
Recombinant Human NGF beta control protein
  • Control Protein
  • PCA Assays
  • ImmunoBlot
Nerve Growth Factor (NGF-β) is a neurotrophic factor related to BDNF, NT-3 and NT-4. NGF-β acts through its receptor β-NGFR, and is involved in the development and maintenance of the sensory and sympathetic nervous systems. NGF-β is also involved in the growth, differentiation and survival of B lymphocytes. Human, mouse and rat proteins show cross-reactivity. Recombinant human NGF-β is a non-covalently linked homodimer, containing two 121 amino acid chains, with a total molecular weight of 27.2 kDa.
Nerve Growth Factor beta Recombinant Protein is suitable as a control for polyclonal or monoclonal anti-Nerve Growth Factor beta in immunological assays.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty